siRNA Details
| Gene Symbol: Lhfpl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172589.2 | NP_766177.1 | 4 | 937 | ||
| GenBank mRNA | AK030103.1 | - | 3 | 1095 | ||
| AK031635.1 | - | 4 | 935 | |||
| AK052969.1 | - | 2 | 799 | |||
| AK087506.1 | - | 4 | 938 | |||
| BC040389.1 | - | 2 | 721 | 3484974 | ||

Lhfpl2
Genomic Map