siRNA Details
| Gene Symbol: Mrps27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173757.3 | NP_776118.2 | 4 | 287 | ||
| GenBank mRNA | AK076008.1 | - | 4 | 286 | ||
| AK087686.1 | - | 4 | 283 | |||
| BC049149.1 | - | 4 | 214 | 6309524 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrps27| Gene Symbol: Mrps27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173757.3 | NP_776118.2 | 4 | 287 | ||
| GenBank mRNA | AK076008.1 | - | 4 | 286 | ||
| AK087686.1 | - | 4 | 283 | |||
| BC049149.1 | - | 4 | 214 | 6309524 | ||