siRNA Details
| Gene Symbol: Sntn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177624.3 | NP_808292.1 | 1, 2 | 134 | ||
| GenBank mRNA | AK040281.1 | - | 1, 2 | 135 | ||
| BC126945.1 | - | 1, 2 | 135 | 40090204 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sntn| Gene Symbol: Sntn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177624.3 | NP_808292.1 | 1, 2 | 134 | ||
| GenBank mRNA | AK040281.1 | - | 1, 2 | 135 | ||
| BC126945.1 | - | 1, 2 | 135 | 40090204 | ||