siRNA Details
| Gene Symbol: Wapal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004436.4 | NP_001004436.2 | 14 | 3418 | ||
| NM_001301330.1 | NP_001288259.1 | 14 | 3400 | |||
| GenBank mRNA | AB167349.1 | - | 14 | 3224 | ||
| AK028167.1 | - | 12 | 1522 | |||
| AK129105.1 | - | 14 | 3418 | |||
| BC037674.1 | - | 8 | 965 | |||
| BC139434.1 | - | 14 | 3173 | 8862456 | ||
| BC145625.1 | - | 14 | 3155 | 9054193 | ||

Wapal
Genomic Map