siRNA Details
| Gene Symbol: Fermt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146054.2 | NP_666166.2 | 5 | 692 | ||
| GenBank mRNA | AK145030.1 | - | 5 | 634 | ||
| AK165608.1 | - | 5 | 662 | |||
| AK166552.1 | - | 5 | 631 | |||
| BC033436.1 | - | 5 | 613 | 4950810 | ||
| BQ179963.1 | - | 4 | 492 | |||
| BU925369.1 | - | 2 | 56 | |||

Fermt2
Genomic Map