siRNA Details
| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172598.3 | NP_766186.2 | 2 | 403 | ||
| GenBank mRNA | AK036390.1 | - | 2 | 362 | ||
| AK042443.1 | - | 1 | 2016 | |||
| AK052690.1 | - | 2 | 379 | |||
| BC063740.1 | - | 2 | 336 | 6328626 | ||

Wdhd1
Genomic Map