siRNA Details
| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172598.3 | NP_766186.2 | 22 | 2967 | ||
| GenBank mRNA | AK036390.1 | - | 22 | 3037 | ||
| AK160777.1 | - | 2 | 128 | |||
| BC063740.1 | - | 22 | 2900 | 6328626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdhd1| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172598.3 | NP_766186.2 | 22 | 2967 | ||
| GenBank mRNA | AK036390.1 | - | 22 | 3037 | ||
| AK160777.1 | - | 2 | 128 | |||
| BC063740.1 | - | 22 | 2900 | 6328626 | ||