siRNA Details
| Gene Symbol: Atg14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172599.4 | NP_766187.1 | 6 | 833 | ||
| GenBank mRNA | AK029330.1 | - | 6 | 814 | ||
| AK029967.1 | - | 6 | 814 | |||
| BC057360.1 | - | 5 | 553 | |||
| BC090995.1 | - | 6 | 813 | 6754513 | ||

Atg14
Genomic Map