siRNA Details
| Gene Symbol: Khnyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027143.2 | NP_081419.1 | 8 | 2160 | ||
| GenBank mRNA | AK032037.1 | - | 8 | 2132 | ||
| AK033704.1 | - | 8 | 2161 | |||
| AK047054.1 | - | 8 | 2706 | |||
| AK047955.1 | - | 8 | 2129 | |||
| AK122247.1 | - | 8 | 2172 | |||
| AK136301.1 | - | 8 | 2188 | |||
| BC116260.1 | - | 8 | 2147 | 40093805 | ||
| BC116261.1 | - | 8 | 2147 | 40093806 | ||

Khnyn
Genomic Map