siRNA Details
| Gene Symbol: Khnyn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027143.2 | NP_081419.1 | 3 | 883 | ||
| GenBank mRNA | AK032037.1 | - | 3 | 855 | ||
| AK033704.1 | - | 3 | 884 | |||
| AK044902.1 | - | 3 | 856 | |||
| AK047054.1 | - | 2 | 1379 | |||
| AK047955.1 | - | 3 | 852 | |||
| AK122247.1 | - | 3 | 835 | |||
| AK136301.1 | - | 3 | 910 | |||
| BC116260.1 | - | 3 | 870 | 40093805 | ||
| BC116261.1 | - | 3 | 870 | 40093806 | ||

Khnyn
Genomic Map