siRNA Details
| Gene Symbol: Spata13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033272.1 | NP_001028444.1 | 9, 10 | 3261 | ||
| GenBank mRNA | AK151394.1 | - | 6, 7 | 1142 | ||
| AK158193.1 | - | 1, 2 | 795 | |||
| AK220313.1 | - | 6, 7 | 983 | |||
| BC035942.1 | - | 7, 8 | 1279 | |||
| BC138455.1 | - | 9, 10 | 3218 | 8861475 | ||
| BC145285.1 | - | 9, 10 | 3218 | 9053839 | ||
| CN538317.1 | - | 3, 4 | 443 | |||

Spata13
Genomic Map