siRNA Details
| Gene Symbol: Fam167a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177628.4 | NP_808296.2 | 3 | 1071 | ||
| GenBank mRNA | AK079469.1 | - | 1 | 147 | ||
| BC060626.1 | - | 1 | 184 | |||
| BC065085.1 | - | 3 | 854 | 6849398 | ||
| BC094561.1 | - | 3 | 981 | |||

Fam167a
Genomic Map