siRNA Details
| Gene Symbol: Ccar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146055.3 | NP_666167.3 | 19 | 2683 | ||
| GenBank mRNA | AK028194.1 | - | 19 | 2603 | ||
| AK031472.1 | - | 15 | 2223 | |||
| BC010499.1 | - | 1 | 1569 | |||
| BC021475.1 | - | 19 | 2713 | 5343456 | ||
| BC025471.1 | - | 6 | 905 | |||
| BC038346.1 | - | 19 | 2635 | 3985250 | ||

Ccar2
Genomic Map