siRNA Details
| Gene Symbol: Ccar2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146055.3 | NP_666167.3 | 8 | 862 | ||
| GenBank mRNA | AK028194.1 | - | 8 | 784 | ||
| AK031472.1 | - | 4 | 402 | |||
| AK050238.1 | - | 8 | 862 | |||
| BC021475.1 | - | 8 | 892 | 5343456 | ||
| BC038346.1 | - | 8 | 814 | 3985250 | ||

Ccar2
Genomic Map