siRNA Details
| Gene Symbol: Vwa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027906.1 | NP_082182.1 | 24 | 2771 | ||
| NM_173758.3 | NP_776119.2 | 24 | 2771 | |||
| GenBank mRNA | AK033642.1 | - | 24 | 2719 | ||
| AK129164.1 | - | 8 | 690 | |||
| AK154108.1 | - | 24 | 2755 | |||

Vwa8
Genomic Map