siRNA Details
| Gene Symbol: Vwa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027906.1 | NP_082182.1 | 12 | 1466 | ||
| NM_173758.3 | NP_776119.2 | 12 | 1466 | |||
| GenBank mRNA | AK004956.1 | - | 12 | 1401 | ||
| AK033642.1 | - | 12 | 1414 | |||
| AK154108.1 | - | 12 | 1450 | |||
| BC019143.1 | - | 12 | 1398 | 5042800 | ||

Vwa8
Genomic Map