siRNA Details
| Gene Symbol: Vwa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027906.1 | NP_082182.1 | 38 | 4730 | ||
| GenBank mRNA | AK030028.1 | - | 2 | 312 | ||
| AK129164.1 | - | 22 | 2649 | |||
| BC037705.1 | - | 3 | 270 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwa8| Gene Symbol: Vwa8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027906.1 | NP_082182.1 | 38 | 4730 | ||
| GenBank mRNA | AK030028.1 | - | 2 | 312 | ||
| AK129164.1 | - | 22 | 2649 | |||
| BC037705.1 | - | 3 | 270 | |||