siRNA Details
| Gene Symbol: Pcdh17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013753.2 | NP_001013775.2 | 1 | 4708 | ||
| GenBank mRNA | AK139275.1 | - | 1 | 2969 | ||
| AK142358.1 | - | 1 | 4704 | |||
| AK147265.1 | - | 1 | 4710 | |||
| AK158668.1 | - | 1 | 2096 | |||
| BC079850.1 | - | 1 | 2705 | |||
| BC089004.1 | - | 1 | 2597 | 6841033 | ||

Pcdh17
Genomic Map