siRNA Details
| Gene Symbol: Farp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134082.3 | NP_598843.3 | 17 | 2237 | ||
| GenBank mRNA | BC030329.1 | - | 10 | 1225 | ||
| DV655111.1 | - | 5 | 598 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Farp1| Gene Symbol: Farp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134082.3 | NP_598843.3 | 17 | 2237 | ||
| GenBank mRNA | BC030329.1 | - | 10 | 1225 | ||
| DV655111.1 | - | 5 | 598 | |||