siRNA Details
| Gene Symbol: Stk24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145465.2 | NP_663440.1 | 4 | 663 | ||
| GenBank mRNA | AK154596.1 | - | 4 | 768 | ||
| AK154959.1 | - | 4 | 664 | |||
| AY188357.1 | - | 4 | 673 | |||
| BC004650.1 | - | 4 | 495 | 3482980 | ||

Stk24
Genomic Map