siRNA Details
Gene Symbol: Timm8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037744.1 | NP_001032833.1 | 2 | 638 | ||
GenBank mRNA | BC099499.1 | - | 2 | 638 | 30931863 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Timm8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037744.1 | NP_001032833.1 | 2 | 638 | ||
GenBank mRNA | BC099499.1 | - | 2 | 638 | 30931863 |