siRNA Details
| Gene Symbol: Ggact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145466.2 | NP_663441.1 | 3 | 762 | ||
| GenBank mRNA | AK050251.1 | - | 3 | 752 | ||
| BB667658.1 | - | 1 | 361 | |||
| BC006662.1 | - | 3 | 732 | 3501534 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggact| Gene Symbol: Ggact | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145466.2 | NP_663441.1 | 3 | 762 | ||
| GenBank mRNA | AK050251.1 | - | 3 | 752 | ||
| BB667658.1 | - | 1 | 361 | |||
| BC006662.1 | - | 3 | 732 | 3501534 | ||