siRNA Details
| Gene Symbol: Itgbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145467.2 | NP_663442.2 | 5 | 885 | ||
| GenBank mRNA | AK029068.1 | - | 5 | 852 | ||
| BC020152.1 | - | 5 | 852 | 3708340 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itgbl1| Gene Symbol: Itgbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145467.2 | NP_663442.2 | 5 | 885 | ||
| GenBank mRNA | AK029068.1 | - | 5 | 852 | ||
| BC020152.1 | - | 5 | 852 | 3708340 | ||