siRNA Details
| Gene Symbol: Trio | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081302.1 | NP_001074771.1 | 47 | 7136 | ||
| GenBank mRNA | BC051169.1 | - | 6 | 603 | ||
| BC060724.1 | - | 27 | 3352 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trio| Gene Symbol: Trio | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081302.1 | NP_001074771.1 | 47 | 7136 | ||
| GenBank mRNA | BC051169.1 | - | 6 | 603 | ||
| BC060724.1 | - | 27 | 3352 | |||