siRNA Details
| Gene Symbol: March6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172606.2 | NP_766194.2 | 19 | 1927 | ||
| GenBank mRNA | AK028353.1 | - | 5 | 461 | ||
| AK042980.1 | - | 4 | 286 | |||
| AK089827.1 | - | 19 | 1929 | |||
| AK129169.1 | - | 7 | 672 | |||
| AK132461.1 | - | 19 | 1900 | |||
| BC037454.1 | - | 5 | 481 | |||
| BC048816.1 | - | 6 | 567 | |||
| BC059190.1 | - | 5 | 455 | |||

March6
Genomic Map