siRNA Details
| Gene Symbol: Eny2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175009.3 | NP_778174.1 | 2 | 177 | ||
| GenBank mRNA | AF173297.1 | - | 2 | 197 | ||
| AK007877.1 | - | 2 | 128 | |||
| AK016562.1 | - | 1 | 1458 | |||
| AK032957.1 | - | 2 | 177 | |||
| AK160628.1 | - | 2 | 144 | |||
| AK162511.1 | - | 2 | 174 | |||
| BC048361.1 | - | 2 | 183 | 4012930 | ||

Eny2
Genomic Map