siRNA Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153548.2 | NP_705776.2 | 11, 12 | 1633 | ||
| GenBank mRNA | AK128970.1 | - | 11, 12 | 1648 | ||
| AK205606.1 | - | 1, 2 | 37 | |||
| BC067035.1 | - | 11, 12 | 1633 | 30532658 | ||

E430025E21Rik
Genomic Map