siRNA Details
| Gene Symbol: Nrbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144847.1 | NP_659096.1 | 8 | 649 | ||
| GenBank mRNA | BC011468.1 | - | 8 | 582 | 4012880 | |
| BC012437.1 | - | 8 | 649 | 3710089 | ||
| BC025042.1 | - | 3 | 111 | |||
| BC068117.1 | - | 13 | 1297 | |||

Nrbp2
Genomic Map