siRNA Details
| Gene Symbol: Arhgap39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168288.1 | NP_001161760.1 | 3, 4 | 760 | ||
| NM_198420.2 | NP_940812.2 | 3, 4 | 760 | |||
| GenBank mRNA | AK035479.1 | - | 2, 3 | 465 | ||
| AK141623.1 | - | 3, 4 | 720 | |||
| AK173232.1 | - | 3, 4 | 695 | |||
| BC060637.1 | - | 3, 4 | 741 | 6849400 | ||

Arhgap39
Genomic Map