siRNA Details
| Gene Symbol: Mkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082536.1 | NP_001076005.1 | 13 | 2818 | ||
| NM_153049.3 | NP_694629.2 | 12 | 2580 | |||
| GenBank mRNA | AF385582.1 | - | 12 | 2351 | ||
| AF532597.1 | - | 13 | 2702 | |||
| AK044188.1 | - | 13 | 2811 | |||
| AK155821.1 | - | 13 | 2824 | |||
| AK155937.1 | - | 12 | 2564 | |||
| BC050941.1 | - | 13 | 2711 | 6821370 | ||
| BC054801.1 | - | 13 | 2754 | |||

Mkl1
Genomic Map