siRNA Details
| Gene Symbol: Mcat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030014.2 | NP_001025185.1 | 3 | 670 | ||
| GenBank mRNA | AK135457.1 | - | 3 | 696 | ||
| AK180925.1 | - | 2 | 207 | |||
| BC025519.1 | - | 1 | 166 | |||
| BC099494.1 | - | 3 | 670 | 30844555 | ||

Mcat
Genomic Map