siRNA Details
| Gene Symbol: Mpped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172610.3 | NP_766198.1 | 3 | 476 | ||
| GenBank mRNA | AK031750.1 | - | 1 | 61 | ||
| AK048421.1 | - | 3 | 477 | |||
| AK158740.1 | - | 3 | 495 | |||
| AK159045.1 | - | 3 | 593 | |||
| AK165755.1 | - | 3 | 437 | |||
| BC057550.1 | - | 2 | 401 | 6810744 | ||

Mpped1
Genomic Map