siRNA Details
| Gene Symbol: Gramd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205353.1 | NP_001192282.1 | 8, 9 | 1013 | ||
| NM_172611.4 | NP_766199.1 | 8, 9 | 915 | |||
| GenBank mRNA | AK036832.1 | - | 8, 9 | 904 | ||
| AK129209.1 | - | 8, 9 | 817 | |||
| AK142266.1 | - | 8, 9 | 868 | |||
| BC021523.1 | - | 4, 5 | 289 | |||
| BC085148.1 | - | 8, 9 | 971 | |||
| BC139228.1 | - | 8, 9 | 904 | 8862250 | ||
| BC139229.1 | - | 8, 9 | 904 | 8862251 | ||

Gramd4
Genomic Map