siRNA Details
| Gene Symbol: Cerk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145475.4 | NP_663450.3 | 11 | 1757 | ||
| GenBank mRNA | AB079067.1 | - | 11 | 1264 | ||
| AK042077.1 | - | 11 | 1348 | |||
| AK052269.1 | - | 11 | 1523 | |||
| AK129416.1 | - | 8 | 899 | |||
| AK161801.1 | - | 595 | ||||
| AK169528.1 | - | 11 | 1342 | |||
| AK213964.1 | - | 1 | 100 | |||
| BC094253.1 | - | 11 | 1756 | 6400872 | ||

Cerk
Genomic Map