siRNA Details
| Gene Symbol: Tbc1d22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145476.2 | NP_663451.2 | 5 | 777 | ||
| GenBank mRNA | AK039627.1 | - | 5 | 790 | ||
| AK146872.1 | - | 5 | 778 | |||
| AK154226.1 | - | 5 | 799 | |||
| AK154602.1 | - | 5 | 820 | |||
| AK155459.1 | - | 5 | 791 | |||
| AK155770.1 | - | 5 | 820 | |||
| BC023106.1 | - | 2 | 172 | 4016550 | ||
| BC066009.1 | - | 5 | 777 | 5708486 | ||

Tbc1d22a
Genomic Map