siRNA Details
| Gene Symbol: Adamts20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177431.4 | NP_803180.3 | 30, 31 | 4878 | ||
| GenBank mRNA | AJ512753.3 | - | 30, 31 | 4647 | ||
| AY189815.1 | - | 30, 31 | 4786 | |||
| BC121792.1 | - | 30, 31 | 4717 | 40099393 | ||

Adamts20
Genomic Map