siRNA Details
| Gene Symbol: Dbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207533.2 | NP_997416.2 | 4 | 1083 | ||
| GenBank mRNA | AK035803.1 | - | 4 | 680 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dbx2| Gene Symbol: Dbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207533.2 | NP_997416.2 | 4 | 1083 | ||
| GenBank mRNA | AK035803.1 | - | 4 | 680 | ||