siRNA Details
| Gene Symbol: Rnd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172612.3 | NP_766200.1 | 4 | 497 | ||
| GenBank mRNA | AK039992.1 | - | 496 | |||
| AK043639.1 | - | 4 | 498 | |||
| AK161870.1 | - | 4 | 494 | |||
| BC048531.1 | - | 4 | 494 | 6535763 | ||

Rnd1
Genomic Map