siRNA Details
| Gene Symbol: Krt73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212485.2 | NP_997650.1 | 5 | 1001 | ||
| GenBank mRNA | BC067067.1 | - | 5 | 1001 | 30538572 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt73| Gene Symbol: Krt73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_212485.2 | NP_997650.1 | 5 | 1001 | ||
| GenBank mRNA | BC067067.1 | - | 5 | 1001 | 30538572 | |