siRNA Details
| Gene Symbol: Krt79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146063.1 | NP_666175.1 | 6 | 1177 | ||
| GenBank mRNA | AK076407.1 | - | 2 | 239 | ||
| BC031593.1 | - | 6 | 1177 | 4935395 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krt79| Gene Symbol: Krt79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146063.1 | NP_666175.1 | 6 | 1177 | ||
| GenBank mRNA | AK076407.1 | - | 2 | 239 | ||
| BC031593.1 | - | 6 | 1177 | 4935395 | ||