siRNA Details
| Gene Symbol: SERPINB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002639.4 | NP_002630.2 | 7 | 1226 | ||
| XM_006722483.2 | XP_006722546.1 | 4 | 734 | |||
| GenBank mRNA | AK301667.1 | - | 6 | 990 | ||
| AK312765.1 | - | 7 | 1204 | |||
| BM955647.1 | - | 2 | 454 | |||
| BX640597.1 | - | 2 | 2622 | |||
| U04313.1 | - | 7 | 1159 | |||

SERPINB5
Genomic Map