siRNA Details
Gene Symbol: SERPINB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002639.4 | NP_002630.2 | 4 | 533 | ||
GenBank mRNA | AK301667.1 | - | 3 | 297 | ||
AK312765.1 | - | 4 | 511 | |||
AL602439.1 | - | 4 | 536 | |||
BC020713.1 | - | 4 | 486 | 4248249 | ||
CV575332.1 | - | 1 | 20 | |||
U04313.1 | - | 4 | 466 |

More Information
Matching Taqman Gene Expression Assay:
Panther Classification:
Gene Ontology Categories:
