siRNA Details
| Gene Symbol: BC048502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177631.3 | NP_808299.1 | 4 | 368 | ||
| GenBank mRNA | AK131692.1 | - | 4 | 380 | ||
| BC048502.1 | - | 4 | 368 | 6740942 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC048502| Gene Symbol: BC048502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177631.3 | NP_808299.1 | 4 | 368 | ||
| GenBank mRNA | AK131692.1 | - | 4 | 380 | ||
| BC048502.1 | - | 4 | 368 | 6740942 | ||