siRNA Details
| Gene Symbol: Cpped1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146067.3 | NP_666179.2 | 2 | 333 | ||
| GenBank mRNA | AK029379.1 | - | 2 | 257 | ||
| AK029442.1 | - | 2 | 356 | |||
| AK032288.1 | - | 2 | 296 | |||
| AK041329.1 | - | 2 | 306 | |||
| AK049721.1 | - | 2 | 334 | |||
| AK080876.1 | - | 2 | 291 | |||
| AK133977.1 | - | 2 | 326 | |||
| AK134718.1 | - | 2 | 334 | |||
| BC025476.1 | - | 2 | 254 | |||

Cpped1
Genomic Map