siRNA Details
| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081154.2 | NP_001074623.1 | 5, 6 | 1413 | ||
| GenBank mRNA | AK035704.1 | - | 1067 | |||
| BC069852.1 | - | 1, 2 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Marf1| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081154.2 | NP_001074623.1 | 5, 6 | 1413 | ||
| GenBank mRNA | AK035704.1 | - | 1067 | |||
| BC069852.1 | - | 1, 2 | 99 | |||