siRNA Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146068.4 | NP_666180.2 | 10 | 1581 | ||
| GenBank mRNA | AK044836.1 | - | 10 | 1564 | ||
| AK052832.1 | - | 10 | 1544 | |||
| AK075871.1 | - | 10 | 1546 | |||
| AK146053.1 | - | 10 | 1581 | |||
| AK166448.1 | - | 5 | 876 | |||
| AK170784.1 | - | 6 | 1213 | |||
| AK172902.1 | - | 10 | 1534 | |||
| BC046558.1 | - | 4 | 585 | |||
| BC069859.1 | - | 1 | 134 | |||
| BC089026.1 | - | 10 | 1518 | 30615298 | ||

Spidr
Genomic Map