siRNA Details
| Gene Symbol: Cyp2ab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183158.3 | NP_898981.2 | 9 | 1687 | ||
| GenBank mRNA | AK089408.1 | - | 9 | 1690 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2ab1| Gene Symbol: Cyp2ab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183158.3 | NP_898981.2 | 9 | 1687 | ||
| GenBank mRNA | AK089408.1 | - | 9 | 1690 | ||