siRNA Details
| Gene Symbol: Atp13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128094.1 | NP_001121566.1 | 5 | 567 | ||
| NM_001128096.1 | NP_001121568.1 | 5 | 567 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp13a3| Gene Symbol: Atp13a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128094.1 | NP_001121566.1 | 5 | 567 | ||
| NM_001128096.1 | NP_001121568.1 | 5 | 567 | |||