siRNA Details
| Gene Symbol: Fam43a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177632.3 | NP_808300.1 | 1 | 1939 | ||
| GenBank mRNA | AK083036.1 | - | 1 | 1939 | ||
| AK137950.1 | - | 1 | 1940 | |||
| BC022623.1 | - | 1 | 532 | |||
| BC026202.1 | - | 2 | 149 | |||
| BC099947.1 | - | 1 | 1952 | 30088416 | ||
| BC138882.1 | - | 1 | 1553 | 8861904 | ||
| BC138884.1 | - | 1 | 1529 | 8861906 | ||

Fam43a
Genomic Map