siRNA Details
| Gene Symbol: Ubxn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177633.4 | NP_808301.3 | 5 | 478 | ||
| GenBank mRNA | AK129218.1 | - | 1 | 76 | ||
| AK147730.1 | - | 5 | 474 | |||
| BC062904.1 | - | 4 | 396 | |||
| CA317987.1 | - | 4 | 396 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubxn7| Gene Symbol: Ubxn7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177633.4 | NP_808301.3 | 5 | 478 | ||
| GenBank mRNA | AK129218.1 | - | 1 | 76 | ||
| AK147730.1 | - | 5 | 474 | |||
| BC062904.1 | - | 4 | 396 | |||
| CA317987.1 | - | 4 | 396 | |||